Learn More
Cayman Chemical 7-methyl-6-Mercaptopurine, Purity: ≥85%, CAS: 3324-79-6, 50 mg
SDP
Catalog No. 502919169
Encompass_Preferred
A derivative of 6-mercaptopurine; has been used as a synthetic intermediate in the synthesis of compounds derived from azathioprine
Catalog No. 50-291-9169 Supplier Cayman Chemical Supplier No. 3965550 MG
A derivative of 6-mercaptopurine; has been used as a synthetic intermediate in the synthesis of compounds derived from azathioprine