Learn More
TARGETMOL CHEMICALS INC 7-METHYL-6-MERCAPTOPURIN 5MG
SDP
Catalog No. 502580699
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 7-Methyl-6-mercaptopurine (7-Methyl-6-thiopurine) is an inhibitor of PRPP aminotransferase inhibits IMP metabolism and prevents purine DNA and RNA synthesis. purity: 99%
Catalog No. 50-258-0699 Supplier TARGETMOL CHEMICALS INC Supplier No. T263995MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 7-Methyl-6-mercaptopurine (7-Methyl-6-thiopurine) is an inhibitor of PRPP aminotransferase inhibits IMP metabolism and prevents purine DNA and RNA synthesis. purity: 99%