Learn More
Cayman Chemical 7-methyl-6-ThIogunosIntec 10mg
SDP
Catalog No. 502970223
Encompass_Preferred
A chromophoric substrate that is used to quantify inorganic phosphate (Pi) in the presence of purine nucleoside phosphorylase, followed by continuous spectrophotometry at an absorbance of 355-360 nm; used to measure changes in Pi levels in coupled enzyme systems
Catalog No. 50-297-0223 Supplier Cayman Chemical Supplier No. 1638110 MG
A chromophoric substrate that is used to quantify inorganic phosphate (Pi) in the presence of purine nucleoside phosphorylase, followed by continuous spectrophotometry at an absorbance of 355-360 nm; used to measure changes in Pi levels in coupled enzyme systems