Learn More
Medchemexpress LLC 7-methyl-DL-tryptophan | 17332-70-6 | MFCD00056926 | 99.5% | 218.25 g/mol | C12H14N2O2 | 250 MG
SDP
Catalog No. 5000282401
Encompass_Preferred
7-Methyl-DL-tryptophan is a methyl-substituted tryptophan analogue used as a biochemical research reagent and synthetic building block. It functions as a precursor in studies of non-ribosomal peptide biosynthesis and is commonly used in biochemical assays. The compound has formula C12H14N2O2 and molecular weight 218.25 g/mol; it is offered in a range of research pack sizes.
Catalog No. 50-002-82401 Supplier Medchemexpress LLC Supplier No. HYW142073250MG
7-Methyl-DL-tryptophan is a methyl-substituted tryptophan analogue used as a biochemical research reagent and synthetic building block. It functions as a precursor in studies of non-ribosomal peptide biosynthesis and is commonly used in biochemical assays. The compound has formula C12H14N2O2 and molecular weight 218.25 g/mol; it is offered in a range of research pack sizes.