Learn More
Cayman Chemical 7-methyl DMT, Purity: ≥98%, CAS: 65882-39-5, 1 mg
SDP
Catalog No. 502978300
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; induces generalization to DOM in drug discrimination tests in rats; intended for research and forensic applications
Catalog No. 50-297-8300 Supplier Cayman Chemical Supplier No. 416841 MG
An analytical reference standard categorized as a tryptamine; induces generalization to DOM in drug discrimination tests in rats; intended for research and forensic applications