Learn More
Medchemexpress LLC 7-Methyl-guanosine-5'-triphosphate sodium | 5 MG
SDP
Catalog No. 5000400676
Encompass_Preferred
7-Methyl-guanosine-5'-triphosphate (m7GTP) sodium is a guanosine 5'-phosphate. Its phosphorothioate analog is a potent cap-dependent translation inhibitor.
  • Potent cap-dependent translation inhibitor.
Catalog No. 50-004-00676 Supplier Medchemexpress LLC Supplier No. HY21586B5MG
7-Methyl-guanosine-5'-triphosphate (m7GTP) sodium is a guanosine 5'-phosphate. Its phosphorothioate analog is a potent cap-dependent translation inhibitor.
  • Potent cap-dependent translation inhibitor.