Learn More
Medchemexpress LLC 7-Methylcoumarin | 2445-83-2 | 50 G
SDP
Catalog No. 5000419496
Encompass_Preferred
7-Methylcoumarin is a coumarin derivative with potent hepatoprotective and antioxidant properties. It acts as a mechanism-based inhibitor for CYP2A6. This compound has been shown to significantly decrease alanine aminotransferase (ALT), aspartate aminotransferase (AST), and serum bilirubin (TB) in rats with CCl4-induced liver damage, while also restoring total protein (TP) and albumin (TA) levels in serum and preventing oxidative stress. Additionally, 7-Methylcoumarin can reduce the mitotic activity of *A. sativum* promeristem.
  • Potent hepatoprotective properties.
  • Antioxidant properties.
  • Mechanism-based inhibitor for CYP2A6.
  • Decreases ALT, AST, and serum bilirubin in rats with CCl4-induced liver damage.
  • Restores total protein and albumin levels in serum.
  • Prevents oxidative stress.
  • Can decline mitotic activity of *A. sativum* promeristem.
Catalog No. 50-004-19496 Supplier Medchemexpress LLC Supplier No. HYN509250G
7-Methylcoumarin is a coumarin derivative with potent hepatoprotective and antioxidant properties. It acts as a mechanism-based inhibitor for CYP2A6. This compound has been shown to significantly decrease alanine aminotransferase (ALT), aspartate aminotransferase (AST), and serum bilirubin (TB) in rats with CCl4-induced liver damage, while also restoring total protein (TP) and albumin (TA) levels in serum and preventing oxidative stress. Additionally, 7-Methylcoumarin can reduce the mitotic activity of *A. sativum* promeristem.
  • Potent hepatoprotective properties.
  • Antioxidant properties.
  • Mechanism-based inhibitor for CYP2A6.
  • Decreases ALT, AST, and serum bilirubin in rats with CCl4-induced liver damage.
  • Restores total protein and albumin levels in serum.
  • Prevents oxidative stress.
  • Can decline mitotic activity of *A. sativum* promeristem.