Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Methylcoumarin | 2445-83-2 | MFCD00006880 | 25 G
SDP

Catalog No. 5000422088
Encompass_Preferred

7-Methylcoumarin is a coumarin derivative with potent hepatoprotective and antioxidant properties. It acts as a mechanism-based inhibitor for CYP2A6, significantly decreasing ALT, AST, and serum bilirubin in rats with CCl4-induced liver damage. It also restores total protein and albumin levels in serum while preventing oxidative stress, and can decline mitotic activity of *A. sativum* promeristem.

  • Coumarin derivative
  • Potent hepatoprotective and antioxidant properties
  • Mechanism-based inhibitor for CYP2A6
  • Decreases ALT, AST, and serum bilirubin in rats with CCl4-induced liver damage
  • Restores total protein and albumin levels in serum
  • Prevents oxidative stress
  • Can decline mitotic activity of *A. sativum* promeristem
  • Purity: 99.34%
  • Appearance: Solid, white to off-white

Catalog No. 50-004-22088 Supplier Medchemexpress LLC Supplier No. 25GHYN5092
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Methylcoumarin is a coumarin derivative with potent hepatoprotective and antioxidant properties. It acts as a mechanism-based inhibitor for CYP2A6, significantly decreasing ALT, AST, and serum bilirubin in rats with CCl4-induced liver damage. It also restores total protein and albumin levels in serum while preventing oxidative stress, and can decline mitotic activity of *A. sativum* promeristem.

  • Coumarin derivative
  • Potent hepatoprotective and antioxidant properties
  • Mechanism-based inhibitor for CYP2A6
  • Decreases ALT, AST, and serum bilirubin in rats with CCl4-induced liver damage
  • Restores total protein and albumin levels in serum
  • Prevents oxidative stress
  • Can decline mitotic activity of *A. sativum* promeristem
  • Purity: 99.34%
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.