Learn More
AdipoGen 7-Methylcoumarin
SDP
Supplier:  AdipoGen CDXM0116G025
Encompass_Preferred
Chemical. CAS 2445-83-2. Formula C10H8O2. MW 160.17. 7-Methylcoumarin is an inhibitor for CYP2A6. It has strong hepatoprotective activity, which could be attributed to its potent antioxidant effects. 7-Methylcoumarin has moderate antimicrobial activity. The compound can be used as internal standard during HPLC analysis.
Catalog No. 50-256-0518