Learn More
AdipoGen 7-Methylguanine
SDP
Supplier:  AdipoGen CDXM0082M250
Encompass_Preferred
Chemical. CAS 578-76-7. Formula C6H7N5O. MW 165.15. Synthetic Integral part of the cap structure of mRNA necessary for the attachment of the nucleic acids to the ribosome. Has binding affinity to guanine deaminase, an important metalloenzyme in purine catabolism. Found in urine of patients with malignant cancer, suggesting an important role.
Catalog No. 50-256-0532