Learn More
Medchemexpress LLC 7-Methylguanine | 578-76-7 | 100.0% | 165.15 | 50 MG
SDP
Catalog No. 5000436446
Encompass_Preferred
7-Methylguanine is an orally active and competitive PARP-1 inhibitor with a Ki value of 61 μM. It is a metabolite of nucleic acids and exhibits anticancer activity against uterine sarcoma and colon adenocarcinoma. It is also used as a probe for protein-DNA interactions.
  • Orally active and competitive PARP-1 inhibitor (Ki = 61 μM)
  • Metabolite of nucleic acids
  • Demonstrates anticancer activity against uterine sarcoma and colon adenocarcinoma
  • Used as a probe for protein-DNA interactions
Catalog No. 50-004-36446 Supplier Medchemexpress LLC Supplier No. HY11335250MG
7-Methylguanine is an orally active and competitive PARP-1 inhibitor with a Ki value of 61 μM. It is a metabolite of nucleic acids and exhibits anticancer activity against uterine sarcoma and colon adenocarcinoma. It is also used as a probe for protein-DNA interactions.
  • Orally active and competitive PARP-1 inhibitor (Ki = 61 μM)
  • Metabolite of nucleic acids
  • Demonstrates anticancer activity against uterine sarcoma and colon adenocarcinoma
  • Used as a probe for protein-DNA interactions