Learn More
Medchemexpress LLC 7-methylguanosine 5'-diphosphate sodium | 104809-16-7 | 97.1% | 479.21 g·mol⁻¹ | C11H16N5NaO11P2 | 5 MG
SDP
Catalog No. 5000222265
Encompass_Preferred
7-Methylguanosine 5'-diphosphate sodium (m7GDP sodium) is a nucleotide cap analog used in RNA and mRNA research. Supplied as a solid sodium salt with high purity, it serves as a reagent or standard in studies of mRNA capping, cap-binding proteins, and enzymatic assays involving guanine nucleotides.
  • Used as a cap analog in mRNA cap-analogue synthesis and biochemical assays.
  • Supplied as a solid sodium salt suitable for laboratory use.
  • High purity (97.1%) for consistent experimental performance.
  • Available in small research pack sizes for low-quantity applications.
  • Store sealed at -20°C; in solution, store at -80°C for longer-term stability.
Catalog No. 50-002-22265 Supplier Medchemexpress LLC Supplier No. HY1414725MG
7-Methylguanosine 5'-diphosphate sodium (m7GDP sodium) is a nucleotide cap analog used in RNA and mRNA research. Supplied as a solid sodium salt with high purity, it serves as a reagent or standard in studies of mRNA capping, cap-binding proteins, and enzymatic assays involving guanine nucleotides.
  • Used as a cap analog in mRNA cap-analogue synthesis and biochemical assays.
  • Supplied as a solid sodium salt suitable for laboratory use.
  • High purity (97.1%) for consistent experimental performance.
  • Available in small research pack sizes for low-quantity applications.
  • Store sealed at -20°C; in solution, store at -80°C for longer-term stability.