Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-methylguanosine-d3 | 00-00-0 | 98.5% | 301.29 g·mol⁻¹ | C11H13D3N5O5 | 5 MG
SDP

Catalog No. 5000264449
Encompass_Preferred

7-Methylguanosine-d3 is a deuterium-labeled stable isotope of 7-methylguanosine intended as a high-purity reference standard for analytical and biochemical applications. It is supplied in small-mass packages suitable for use in quantitative mass spectrometry, tracer studies, and method development. The unlabeled parent compound has reported activity against cNIIIB nucleotidase (IC50 = 87.8 ± 7.5 μM).

  • High-purity deuterium-labeled isotopic standard for accurate mass spectrometry.
  • Suitable as an internal standard for LC-MS or GC-MS quantitation.
  • Useful for tracer studies and metabolic labeling experiments.
  • Provided in small mass vials compatible with analytical workflows.
  • Molecular formula C11H13D3N5O5; molecular weight 301.29 g·mol⁻¹.

Catalog No. 50-002-64449 Supplier Medchemexpress LLC Supplier No. HY122524S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

7-Methylguanosine-d3 is a deuterium-labeled stable isotope of 7-methylguanosine intended as a high-purity reference standard for analytical and biochemical applications. It is supplied in small-mass packages suitable for use in quantitative mass spectrometry, tracer studies, and method development. The unlabeled parent compound has reported activity against cNIIIB nucleotidase (IC50 = 87.8 ± 7.5 μM).

  • High-purity deuterium-labeled isotopic standard for accurate mass spectrometry.
  • Suitable as an internal standard for LC-MS or GC-MS quantitation.
  • Useful for tracer studies and metabolic labeling experiments.
  • Provided in small mass vials compatible with analytical workflows.
  • Molecular formula C11H13D3N5O5; molecular weight 301.29 g·mol⁻¹.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.