Learn More
Medchemexpress LLC 7-Methylguanosine-d3 | 20244-86-4 | 98.5% | 301.29 | 1 MG
SDP
Catalog No. 5000438592
Encompass_Preferred
7-Methylguanosine-d3 is a deuterium-labeled variant of 7-Methylguanosine, which functions as a cNIIIB nucleotidase inhibitor with an IC50 value of 87.8 ± 7.5 μM. Deuterium-labeled compounds are frequently used in drug development as tracers for quantitative analysis, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Can be an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution may affect pharmacokinetic and metabolic profiles.
Catalog No. 50-004-38592 Supplier Medchemexpress LLC Supplier No. HY122524S1MG
7-Methylguanosine-d3 is a deuterium-labeled variant of 7-Methylguanosine, which functions as a cNIIIB nucleotidase inhibitor with an IC50 value of 87.8 ± 7.5 μM. Deuterium-labeled compounds are frequently used in drug development as tracers for quantitative analysis, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Can be an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution may affect pharmacokinetic and metabolic profiles.