Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-methylguanosine iodide | 81100-62-1 | 98.5% | 10 MG
SDP

Catalog No. 5000232161
Encompass_Preferred

7-Methylguanosine iodide is the iodide salt of the modified nucleoside 7-methylguanosine. It is supplied for use as an analytical standard and research reagent in studies of RNA cap structure and nucleoside metabolism. Refer to the product certificate of analysis for purity and storage instructions.

  • Iodide salt of 7-methylguanosine.
  • Intended for analytical and research use related to RNA cap structure and nucleoside metabolism.
  • Molecular formula C11H16IN5O5 and molecular weight 425.18 g/mol.
  • CAS number 81100-62-1 for substance identification.
  • Typically supplied as a solid; check the certificate of analysis for storage and stability.
  • Packaged in 10 mg quantities for analytical or small-scale use.

Catalog No. 50-002-32161 Supplier Medchemexpress LLC Supplier No. HY122524A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Methylguanosine iodide is the iodide salt of the modified nucleoside 7-methylguanosine. It is supplied for use as an analytical standard and research reagent in studies of RNA cap structure and nucleoside metabolism. Refer to the product certificate of analysis for purity and storage instructions.

  • Iodide salt of 7-methylguanosine.
  • Intended for analytical and research use related to RNA cap structure and nucleoside metabolism.
  • Molecular formula C11H16IN5O5 and molecular weight 425.18 g/mol.
  • CAS number 81100-62-1 for substance identification.
  • Typically supplied as a solid; check the certificate of analysis for storage and stability.
  • Packaged in 10 mg quantities for analytical or small-scale use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.