Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-methylguanosine iodide | 81100-62-1 | 99.6% | 25 MG
SDP

Catalog No. 5000230968
Encompass_Preferred

7-Methylguanosine iodide is the iodide salt of 7-methylguanosine, a modified nucleoside used as a research reagent and analytical standard. It is relevant to studies of RNA cap structures and nucleoside chemistry.

  • Modified nucleoside present in various RNAs.
  • Used as a research reagent and analytical standard.
  • CAS number 81100-62-1.
  • Molecular formula C11H16IN5O5 and molecular weight 425.18 g/mol.
  • Supplied as small pre-weighed laboratory packs for analytical use.

Catalog No. 50-002-30968 Supplier Medchemexpress LLC Supplier No. HY122524A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Methylguanosine iodide is the iodide salt of 7-methylguanosine, a modified nucleoside used as a research reagent and analytical standard. It is relevant to studies of RNA cap structures and nucleoside chemistry.

  • Modified nucleoside present in various RNAs.
  • Used as a research reagent and analytical standard.
  • CAS number 81100-62-1.
  • Molecular formula C11H16IN5O5 and molecular weight 425.18 g/mol.
  • Supplied as small pre-weighed laboratory packs for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.