Learn More
Cayman Chemical 7-Methylguanine, Purity: ≥98%, CAS: 578-76-7, 100 mg
SDP
Catalog No. 502966119
Encompass_Preferred
A metabolite of the natural purine base that is generated by DNA methylation and has been used as a probe of protein-DNA interactions and for DNA sequencing methods; carcinogenic biomarker for cigarette smoking that is detectable in urine
Catalog No. 50-296-6119 Supplier Cayman Chemical Supplier No. 18161100 MG
A metabolite of the natural purine base that is generated by DNA methylation and has been used as a probe of protein-DNA interactions and for DNA sequencing methods; carcinogenic biomarker for cigarette smoking that is detectable in urine