Learn More
Cayman Chemical 7-MethylgunIn 50mg
SDP
Catalog No. 502965528
Encompass_Preferred
A metabolite of the natural purine base that is generated by DNA methylation and has been used as a probe of protein-DNA interactions and for DNA sequencing methods; carcinogenic biomarker for cigarette smoking that is detectable in urine
Catalog No. 50-296-5528 Supplier Cayman Chemical Supplier No. 1816150 MG
A metabolite of the natural purine base that is generated by DNA methylation and has been used as a probe of protein-DNA interactions and for DNA sequencing methods; carcinogenic biomarker for cigarette smoking that is detectable in urine