Learn More
Cayman Chemical 7-Methylguanosine, Purity: ≥98%, CAS: 20244-86-4, 100 mg
SDP
Catalog No. 502969216
Encompass_Preferred
A methylated form of guanosine; an RNA modification that is present in the 5’-terminal cap of mRNA, as well as in tRNA; has been used as a substrate to measure the activity of PNP; urinary levels are decreased in patients with prostate cancer
Catalog No. 50-296-9216 Supplier Cayman Chemical Supplier No. 15988100 MG
A methylated form of guanosine; an RNA modification that is present in the 5’-terminal cap of mRNA, as well as in tRNA; has been used as a substrate to measure the activity of PNP; urinary levels are decreased in patients with prostate cancer