Learn More
Medchemexpress LLC 7-Methylindole | 933-67-5 | 131.17 | 25 G
SDP
Catalog No. 5000449190
Encompass_Preferred
7-Methylindole is a biochemical reagent suitable for life science research as a biological material or organic compound. It is for research use only.
  • Reactant for the preparation of pharmaceutically active pyrrolidine analogues via Markovnikov addition
  • Reactant for tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Reactant for plant growth inhibitors
  • Reactant for antifungal agents
  • Reactant for sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes
  • Reactant for inhibitors of HIV-1 attachment
  • Reactant for Friedel-Crafts alkylation reactions and reductive formylation reactions
Catalog No. 50-004-49190 Supplier Medchemexpress LLC Supplier No. 25GHYW007670
7-Methylindole is a biochemical reagent suitable for life science research as a biological material or organic compound. It is for research use only.
  • Reactant for the preparation of pharmaceutically active pyrrolidine analogues via Markovnikov addition
  • Reactant for tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Reactant for plant growth inhibitors
  • Reactant for antifungal agents
  • Reactant for sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes
  • Reactant for inhibitors of HIV-1 attachment
  • Reactant for Friedel-Crafts alkylation reactions and reductive formylation reactions