Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Methylisatin | 1127-59-9 | 161.16 | 10 G
SDP

Catalog No. 5000446509
Encompass_Preferred

7-Methylisatin is a derivative of Isatin and can be utilized as a molecular block for biochemical assay reagents.

  • Derivative of Isatin
  • Can be utilized as a molecular block
  • For research use only
  • Store at room temperature for 3 years; in solvent, -80°C for 2 years; -20°C for 1 year
  • Soluble in DMSO

Catalog No. 50-004-46509 Supplier Medchemexpress LLC Supplier No. HYW00771910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Methylisatin is a derivative of Isatin and can be utilized as a molecular block for biochemical assay reagents.

  • Derivative of Isatin
  • Can be utilized as a molecular block
  • For research use only
  • Store at room temperature for 3 years; in solvent, -80°C for 2 years; -20°C for 1 year
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.