Learn More
Medchemexpress LLC HY-W017163 50mg Medchemexpress, 7-Methylxanthine CAS:552-62-5 Purity:>98%
SDP
Catalog No. 501934629
Encompass_Preferred
Medchemexpress, HY-W017163 50mg 7-Methylxanthine CAS:552-62-5 7-Methylxanthine, a methyl derivative of xanthine, is one of the purine components in urinary calculi. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4629 Supplier Medchemexpress LLC Supplier No. HYW01716350MG
Medchemexpress, HY-W017163 50mg 7-Methylxanthine CAS:552-62-5 7-Methylxanthine, a methyl derivative of xanthine, is one of the purine components in urinary calculi. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.