Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-methylxanthine | 552-62-5 | MFCD00037979 | >=98.0% | 166.14 g/mol | C6H6N4O2 | 1 ML
SDP

Catalog No. 5000300926
Encompass_Preferred

7-methylxanthine is a methylated xanthine derivative used in biochemical and pharmacological research as an adenosine receptor antagonist and endogenous metabolite. Supplied for research and analytical applications, it has formula C6H6N4O2 and a molecular weight of 166.14 g/mol. Purity is commonly reported as >=98.0% (HPLC); consult the certificate of analysis for batch-specific purity.

  • Adenosine receptor antagonist and endogenous metabolite for receptor and metabolism studies.
  • Chemical identity: CAS 552-62-5, formula C6H6N4O2, molecular weight 166.14 g/mol.
  • Available as solid formats and can be prepared or provided as a 10 mM solution in DMSO.
  • Typical purity reported as >=98.0% (HPLC); verify with certificate of analysis.
  • Intended for research and analytical applications; not for human or clinical use.

Catalog No. 50-003-00926 Supplier Medchemexpress LLC Supplier No. HYW01716310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-methylxanthine is a methylated xanthine derivative used in biochemical and pharmacological research as an adenosine receptor antagonist and endogenous metabolite. Supplied for research and analytical applications, it has formula C6H6N4O2 and a molecular weight of 166.14 g/mol. Purity is commonly reported as >=98.0% (HPLC); consult the certificate of analysis for batch-specific purity.

  • Adenosine receptor antagonist and endogenous metabolite for receptor and metabolism studies.
  • Chemical identity: CAS 552-62-5, formula C6H6N4O2, molecular weight 166.14 g/mol.
  • Available as solid formats and can be prepared or provided as a 10 mM solution in DMSO.
  • Typical purity reported as >=98.0% (HPLC); verify with certificate of analysis.
  • Intended for research and analytical applications; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.