Learn More
Medchemexpress LLC 7-methylxanthine | 552-62-5 | MFCD00037979 | 99.9% | 166.14 g·mol⁻¹ | C6H6N4O2 | 100 MG
SDP
Catalog No. 5000301162
Encompass_Preferred
7-Methylxanthine is a methylxanthine derivative used as a research tool and non-selective adenosine receptor antagonist. It is supplied as a solid for biochemical and pharmacological studies involving adenosine receptor signaling.
  • High purity (~99.9%) suitable for analytical and biological assays.
  • Well-characterized chemical with CAS number 552-62-5 for easy identification.
  • Solid form, white to light brown, convenient for storage and handling.
  • Compatible with common solvents such as DMSO for solution preparations.
  • Recommended storage at low temperature to maintain stability.
Catalog No. 50-003-01162 Supplier Medchemexpress LLC Supplier No. HYW017163100MG
7-Methylxanthine is a methylxanthine derivative used as a research tool and non-selective adenosine receptor antagonist. It is supplied as a solid for biochemical and pharmacological studies involving adenosine receptor signaling.
  • High purity (~99.9%) suitable for analytical and biological assays.
  • Well-characterized chemical with CAS number 552-62-5 for easy identification.
  • Solid form, white to light brown, convenient for storage and handling.
  • Compatible with common solvents such as DMSO for solution preparations.
  • Recommended storage at low temperature to maintain stability.