Learn More
Medchemexpress LLC 7-methylxanthine-d3 | 1185878-99-2 | 99.3% | 169.16 | C6H3D3N4O2 | 1 MG
SDP
Catalog No. 5000301860
Encompass_Preferred
7-Methylxanthine-d3 is a deuterium-labeled isotopic standard of 7-methylxanthine supplied as a high-purity solid for research use. The d3 label provides a predictable mass shift that enables clear differentiation from the unlabeled analyte in mass spectrometry and tracer experiments. Molecular formula: C6H3D3N4O2; molecular weight: 169.16; CAS: 1185878-99-2.
  • High purity 99.31% for analytical reliability.
  • Deuterium-labeled (d3) for mass-shifted internal standard use.
  • Suitable for LC-MS/MS quantitation and tracer studies.
  • Available in small package sizes (1 mg, 5 mg, 10 mg) for method development.
  • Intended for research use only.
Catalog No. 50-003-01860 Supplier Medchemexpress LLC Supplier No. HYW017163S11MG
7-Methylxanthine-d3 is a deuterium-labeled isotopic standard of 7-methylxanthine supplied as a high-purity solid for research use. The d3 label provides a predictable mass shift that enables clear differentiation from the unlabeled analyte in mass spectrometry and tracer experiments. Molecular formula: C6H3D3N4O2; molecular weight: 169.16; CAS: 1185878-99-2.
  • High purity 99.31% for analytical reliability.
  • Deuterium-labeled (d3) for mass-shifted internal standard use.
  • Suitable for LC-MS/MS quantitation and tracer studies.
  • Available in small package sizes (1 mg, 5 mg, 10 mg) for method development.
  • Intended for research use only.