Learn More
Cayman Chemical 7-MethylxnthIn 100mg
SDP
Catalog No. 502947323
Encompass_Preferred
An adenosine A1 receptor antagonist and active metabolite of theobromine and caffeine; inhibits binding of cyclohexyladensoine in rat cerebral cortical membranes; inhibits eye elongation and reduces decreases in the diameter of scleral collagen fibrils in a guinea pig model of form-deprivation myopia at 300 mg/kg
Catalog No. 50-294-7323 Supplier Cayman Chemical Supplier No. 34348100 MG
An adenosine A1 receptor antagonist and active metabolite of theobromine and caffeine; inhibits binding of cyclohexyladensoine in rat cerebral cortical membranes; inhibits eye elongation and reduces decreases in the diameter of scleral collagen fibrils in a guinea pig model of form-deprivation myopia at 300 mg/kg