Learn More
Cayman Chemical 7-Methylxanthine, Purity: ≥98%, CAS: 552-62-5, 250 mg
SDP
Catalog No. 502948383
Encompass_Preferred
An adenosine A1 receptor antagonist and active metabolite of theobromine and caffeine; inhibits binding of cyclohexyladensoine in rat cerebral cortical membranes; inhibits eye elongation and reduces decreases in the diameter of scleral collagen fibrils in a guinea pig model of form-deprivation myopia at 300 mg/kg
Catalog No. 50-294-8383 Supplier Cayman Chemical Supplier No. 34348250 MG
An adenosine A1 receptor antagonist and active metabolite of theobromine and caffeine; inhibits binding of cyclohexyladensoine in rat cerebral cortical membranes; inhibits eye elongation and reduces decreases in the diameter of scleral collagen fibrils in a guinea pig model of form-deprivation myopia at 300 mg/kg