Learn More
Cayman Chemical 7-Methylxanthine, Purity: ≥98%, CAS: 552-62-5, 500 mg
SDP
Catalog No. 502947505
Encompass_Preferred
An adenosine A1 receptor antagonist and active metabolite of theobromine and caffeine; inhibits binding of cyclohexyladensoine in rat cerebral cortical membranes; inhibits eye elongation and reduces decreases in the diameter of scleral collagen fibrils in a guinea pig model of form-deprivation myopia at 300 mg/kg
Catalog No. 50-294-7505 Supplier Cayman Chemical Supplier No. 34348500 MG
An adenosine A1 receptor antagonist and active metabolite of theobromine and caffeine; inhibits binding of cyclohexyladensoine in rat cerebral cortical membranes; inhibits eye elongation and reduces decreases in the diameter of scleral collagen fibrils in a guinea pig model of form-deprivation myopia at 300 mg/kg