Learn More
USA Scientific Inc 7 mL transfer pipet, graduated
Catalog No. 502745168
Encompass
7 mL polyethylene transfer pipet, graduated to 3 ml, bulb draw 3.5 ml, length 16.0 cm. 21 Drops per mL, bulk, 250/pack.
Catalog No. 50-274-5168 Supplier USA Scientific Inc Supplier No. 10201400
7 mL polyethylene transfer pipet, graduated to 3 ml, bulb draw 3.5 ml, length 16.0 cm. 21 Drops per mL, bulk, 250/pack.