Learn More
Medchemexpress LLC 7-(morpholin-4-yl)-2,1,3-benzoxadiazol-4-amine | 842964-18-5 | 99.9% | 220.23 g/mol | C10H12N4O2 | 25MG
SDP
Catalog No. 5000236504
Encompass_Preferred
δ-Secretase inhibitor 11 is an orally active, blood-brain barrier-penetrant, selective δ-secretase inhibitor (IC50 = 0.7 μM) developed for Alzheimer's disease research; it reduces tau and APP cleavage and has demonstrated improvement of synaptic function in transgenic mouse models. The compound is supplied as a solid with high research-grade purity and is soluble in DMSO for in vitro use.
  • Orally active and blood-brain barrier penetrant.
  • Selective δ-secretase inhibition with reported IC50 of 0.7 μM.
  • High chemical purity suitable for research (99.9%).
  • Soluble in DMSO (~116.7 mg/mL) with ultrasonic assistance.
  • Solid form; store at 4°C and protect from light.
Catalog No. 50-002-36504 Supplier Medchemexpress LLC Supplier No. HY12483225MG
Add to Cart
Add to Cart
δ-Secretase inhibitor 11 is an orally active, blood-brain barrier-penetrant, selective δ-secretase inhibitor (IC50 = 0.7 μM) developed for Alzheimer's disease research; it reduces tau and APP cleavage and has demonstrated improvement of synaptic function in transgenic mouse models. The compound is supplied as a solid with high research-grade purity and is soluble in DMSO for in vitro use.
  • Orally active and blood-brain barrier penetrant.
  • Selective δ-secretase inhibition with reported IC50 of 0.7 μM.
  • High chemical purity suitable for research (99.9%).
  • Soluble in DMSO (~116.7 mg/mL) with ultrasonic assistance.
  • Solid form; store at 4°C and protect from light.