Learn More
Medchemexpress LLC 7-(morpholin-4-yl)-2,1,3-benzoxadiazol-4-amine | 842964-18-5 | MFCD03422529 | 99.9% | 220.23 g/mol | C10H12N4O2 | 10MG
SDP
Catalog No. 5000239477
Encompass_Preferred
δ-Secretase inhibitor 11 is a selective, orally active small-molecule inhibitor of δ-secretase used in neurodegeneration research. It is reported to be blood-brain barrier permeable and non-toxic in preclinical models, attenuating pathological cleavage of tau and amyloid precursor protein and improving synaptic function and cognition in relevant transgenic mouse models.
  • Selective δ-secretase inhibition with reported IC50 0.7 μM.
  • Blood-brain barrier permeable, suitable for in vivo studies.
  • Reduces tau and APP cleavage in disease models.
  • High reported purity (≈99.9%).
  • Available as solid quantities and as 10 mM solution in DMSO.
  • Useful for Alzheimer's disease mechanism and therapeutic research.
Catalog No. 50-002-39477 Supplier Medchemexpress LLC Supplier No. HY12483210MG
δ-Secretase inhibitor 11 is a selective, orally active small-molecule inhibitor of δ-secretase used in neurodegeneration research. It is reported to be blood-brain barrier permeable and non-toxic in preclinical models, attenuating pathological cleavage of tau and amyloid precursor protein and improving synaptic function and cognition in relevant transgenic mouse models.
  • Selective δ-secretase inhibition with reported IC50 0.7 μM.
  • Blood-brain barrier permeable, suitable for in vivo studies.
  • Reduces tau and APP cleavage in disease models.
  • High reported purity (≈99.9%).
  • Available as solid quantities and as 10 mM solution in DMSO.
  • Useful for Alzheimer's disease mechanism and therapeutic research.