Learn More
Medchemexpress LLC 7-morpholin-4-yl-benzo[1,2,5]oxadiazol-4-ylamine | 842964-18-5 | MFCD03422529 | 98.0% | 220.23 g/mol | C10H12N4O2 | 10MM 1ML
SDP
Catalog No. 5000237789
Encompass_Preferred
Delta-Secretase Inhibitor 11 (CAS 842964-18-5) is a small-molecule research compound that inhibits δ-secretase activity. It is supplied as a 10 mM solution in a 1 mL vial and is used as a biochemical tool to probe delta-secretase-related pathways. The compound has molecular formula C10H12N4O2 and a molecular weight of 220.23 g/mol.
Catalog No. 50-002-37789 Supplier Medchemexpress LLC Supplier No. HY12483210MM 1ML
Delta-Secretase Inhibitor 11 (CAS 842964-18-5) is a small-molecule research compound that inhibits δ-secretase activity. It is supplied as a 10 mM solution in a 1 mL vial and is used as a biochemical tool to probe delta-secretase-related pathways. The compound has molecular formula C10H12N4O2 and a molecular weight of 220.23 g/mol.