Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-nitro-2,1,3-benzoxadiazol-4-ylamine | 10199-91-4 | 96.7% | 10 MG
SDP

Catalog No. 5000233423
Encompass_Preferred

NBD-amine (7-nitro-2,1,3-benzoxadiazol-4-ylamine) is a fluorogenic amine used as a fluorescent probe and a synthetic building block for labeling and sensing applications. It fluoresces weakly in water and strongly in organic solvents, membranes, and other hydrophobic environments. CAS 10199-91-4; purity ~96.7%.

  • Fluorogenic amine that enhances fluorescence in hydrophobic environments.
  • Useful for labeling, sensing, and probe development.
  • Soluble in DMSO at 31.25 mg/mL; ultrasonic agitation may be needed.
  • Powder stable at -20°C for up to 3 years.
  • Molecular weight 180.12 g/mol; formula C6H4N4O3.

Catalog No. 50-002-33423 Supplier Medchemexpress LLC Supplier No. HY11979110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NBD-amine (7-nitro-2,1,3-benzoxadiazol-4-ylamine) is a fluorogenic amine used as a fluorescent probe and a synthetic building block for labeling and sensing applications. It fluoresces weakly in water and strongly in organic solvents, membranes, and other hydrophobic environments. CAS 10199-91-4; purity ~96.7%.

  • Fluorogenic amine that enhances fluorescence in hydrophobic environments.
  • Useful for labeling, sensing, and probe development.
  • Soluble in DMSO at 31.25 mg/mL; ultrasonic agitation may be needed.
  • Powder stable at -20°C for up to 3 years.
  • Molecular weight 180.12 g/mol; formula C6H4N4O3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.