Learn More
Cayman Chemical 7-Nitroindazole, Purity: ≥98%, CAS: 2942-42-9, 100 mg
SDP
Catalog No. 502894074
Encompass_Preferred
A non-selective inhibitor of NOS isoforms in vitro (IC50s = 0.71, 0.78, and 5.8 µM for rat nNOS, bovine eNOS, and rat iNOS, respectively); shows good anti-nociceptive effects without affecting blood pressure via inhibition of eNOS in vivo
Catalog No. 50-289-4074 Supplier Cayman Chemical Supplier No. 81340100 MG
A non-selective inhibitor of NOS isoforms in vitro (IC50s = 0.71, 0.78, and 5.8 µM for rat nNOS, bovine eNOS, and rat iNOS, respectively); shows good anti-nociceptive effects without affecting blood pressure via inhibition of eNOS in vivo