Learn More
Medchemexpress LLC 7-Nitroindazole | 2942-42-9 | 99.1% | C7H5N3O2 | 500 MG
SDP
Catalog No. 5000434130
Encompass_Preferred
7-Nitroindazole is a selective and BBB-penetrable inhibitor of nitric oxide synthase (NOS) with anti-injurious and neuroprotective properties, making it useful for research into neurological diseases such as Parkinson's disease.
  • Selective and BBB-penetrable inhibitor of nitric oxide synthase (NOS).
  • Inhibits nNOS in experimental animals without affecting blood pressure or endothelium-dependent relaxation.
  • Exhibits anti-injurious and anti-nociceptive effects in a formalin-induced mouse pain model.
  • Improves L-DOPA-induced dyskinesias in non-human primate models of Parkinson's disease.
  • Enhances learning and memory in hypothyroid rats.
  • Potentiates c-fos expression induced by muscle proprioceptive signaling in the spinal cord.
Catalog No. 50-004-34130 Supplier Medchemexpress LLC Supplier No. HY69019500MG
7-Nitroindazole is a selective and BBB-penetrable inhibitor of nitric oxide synthase (NOS) with anti-injurious and neuroprotective properties, making it useful for research into neurological diseases such as Parkinson's disease.
  • Selective and BBB-penetrable inhibitor of nitric oxide synthase (NOS).
  • Inhibits nNOS in experimental animals without affecting blood pressure or endothelium-dependent relaxation.
  • Exhibits anti-injurious and anti-nociceptive effects in a formalin-induced mouse pain model.
  • Improves L-DOPA-induced dyskinesias in non-human primate models of Parkinson's disease.
  • Enhances learning and memory in hypothyroid rats.
  • Potentiates c-fos expression induced by muscle proprioceptive signaling in the spinal cord.