Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Nitroindole | 6960-42-5 | 162.15 | 10 G
SDP

Catalog No. 5000445956
Encompass_Preferred

7-Nitroindole is a cleavable base analog and a photochemical precursor. It is used to specifically generate 2'-deoxyribonolactone oxidative damage sites in DNA strands and for the study of photo-controlled DNA breakage tools.

  • Solid appearance with color ranging from yellow to green
  • Soluble in DMSO at 100 mg/mL
  • Molecular formula: C8H6N2O2
  • Targets DNA/RNA synthesis
  • Pathway: Cell Cycle/DNA Damage

Catalog No. 50-004-45956 Supplier Medchemexpress LLC Supplier No. HYW00197410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

7-Nitroindole is a cleavable base analog and a photochemical precursor. It is used to specifically generate 2'-deoxyribonolactone oxidative damage sites in DNA strands and for the study of photo-controlled DNA breakage tools.

  • Solid appearance with color ranging from yellow to green
  • Soluble in DMSO at 100 mg/mL
  • Molecular formula: C8H6N2O2
  • Targets DNA/RNA synthesis
  • Pathway: Cell Cycle/DNA Damage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.