Learn More
Medchemexpress LLC 7-Nitroindole | 6960-42-5 | 162.15 | 5 G
SDP
Catalog No. 5000447690
Encompass_Preferred
7-Nitroindole is a cleavable base analog, serving as a photochemical precursor used to specifically generate 2'-deoxyribonolactone oxidative damage sites in DNA strands. It can also be utilized for the study of photo-controlled DNA breakage tools.
  • Cleavable base analog
  • Photochemical precursor for 2'-deoxyribonolactone oxidative damage sites
  • Used in the study of photo-controlled DNA breakage tools
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-47690 Supplier Medchemexpress LLC Supplier No. HYW0019745G
7-Nitroindole is a cleavable base analog, serving as a photochemical precursor used to specifically generate 2'-deoxyribonolactone oxidative damage sites in DNA strands. It can also be utilized for the study of photo-controlled DNA breakage tools.
  • Cleavable base analog
  • Photochemical precursor for 2'-deoxyribonolactone oxidative damage sites
  • Used in the study of photo-controlled DNA breakage tools
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month