Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Nitroindole 98.0+%, TCI America™
SDP

Catalog No. N06855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06855G 5 g
N06851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06855G Supplier TCI America Supplier No. N06855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6960-42-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00005683
InChI Key LZJGQIVWUKFTRD-UHFFFAOYSA-N
Synonym 7-nitroindole, 1h-indole, 7-nitro, indole, 7-nitro, 7-nitro indole, pubchem7441, acmc-1b2jq, 5-20-07-00044 beilstein handbook reference, ksc353k7l, 1h-indole, 7-nitro-9ci
PubChem CID 23396
IUPAC Name 7-nitro-1H-indole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2

Specifications

Melting Point 97°C
Color Yellow
Quantity 5 g
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Nitroindole
RTECSNumber NM1169000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.