Learn More
Cayman Chemical 7-nor-7-carboxy CnabIdI 1mg
SDP
Catalog No. 502987581
Encompass_Preferred
An analytical reference standard categorized as a phytocannabinoid metabolite; a metabolite of cannabidiol; intended for research and forensic applications
Catalog No. 50-298-7581 Supplier Cayman Chemical Supplier No. 216371 MG
An analytical reference standard categorized as a phytocannabinoid metabolite; a metabolite of cannabidiol; intended for research and forensic applications