Learn More
Medchemexpress LLC HY-141147 25mg Medchemexpress, 7-O-(Amino-PEG4)-paclitaxel CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14114725MG
Encompass_Preferred
Medchemexpress, HY-141147 25mg 7-O-(Amino-PEG4)-paclitaxel CAS: 7-O-(Amino-PEG4)-paclitaxel is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1559