Learn More
Medchemexpress LLC 7-O-Ethylmorroniside | 945721-10-8 | 5 MG
SDP
Catalog No. 5000421514
Encompass_Preferred
7-O-Ethylmorroniside is an iridoid glucoside isolated from the fruit of *Cornus officinalis*, which is traditionally used in Chinese medicine. This compound is primarily researched for its potential in treating kidney diseases, including diabetic nephropathy.
  • Iridoid glucoside
  • Isolated from the fruit of *Cornus officinalis*
  • High purity of 99.91% (HPLC)
  • Researched for kidney diseases, including diabetic nephropathy
Catalog No. 50-004-21514 Supplier Medchemexpress LLC Supplier No. HYN26085MG
7-O-Ethylmorroniside is an iridoid glucoside isolated from the fruit of *Cornus officinalis*, which is traditionally used in Chinese medicine. This compound is primarily researched for its potential in treating kidney diseases, including diabetic nephropathy.
  • Iridoid glucoside
  • Isolated from the fruit of *Cornus officinalis*
  • High purity of 99.91% (HPLC)
  • Researched for kidney diseases, including diabetic nephropathy