Learn More
Medchemexpress LLC HY-N2746 1mg Medchemexpress, 7-O-Geranylscopoletin CAS:28587-43-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN27461MG
Encompass_Preferred
Medchemexpress, HY-N2746 1mg 7-O-Geranylscopoletin CAS:28587-43-1 7-O-Geranylscopoletin is a coumarin from the root of Atalantia monophylla. Various parts of this plant have been used for folk medicine for several purposes such as the treatment of chronic rheumatism, paralysis, antispasmodic, stimulant and hemiplegia [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3934