Learn More
AdipoGen 7-O-Methylaloeresin A (1 mg)
SDP
Supplier:  AdipoGen AGCN20491M001
Encompass_Preferred
Chemical. CAS: 329361-25-3. Formula: C29H30O11. MW: 554.5. Isolated from Aloe vera. Metabolite of aloesin. Potent antioxidant (DPPH assay). Antibiotic. Antifungal. Moderate BACE inhibitor.
Catalog No. 50-205-3675