Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Octyn-1-ol | 871-91-0 | 99.6% | 126.20 | 1 G
SDP

Catalog No. 5000422368
Encompass_Preferred

7-Octyn-1-ol is a precursor to 7-Octynoic acid, formed through its oxidation. It functions as a click chemistry reagent, containing an alkyne group that enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing azide groups.

  • Precursor to 7-Octynoic acid
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules possessing azide groups
  • Molecular formula: C8H14O
  • Molecular weight: 126.20
  • Appearance: Colorless to light yellow liquid

Catalog No. 50-004-22368 Supplier Medchemexpress LLC Supplier No. HYW0085541G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Octyn-1-ol is a precursor to 7-Octynoic acid, formed through its oxidation. It functions as a click chemistry reagent, containing an alkyne group that enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing azide groups.

  • Precursor to 7-Octynoic acid
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules possessing azide groups
  • Molecular formula: C8H14O
  • Molecular weight: 126.20
  • Appearance: Colorless to light yellow liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.