Learn More
Medchemexpress LLC 7-Octyn-1-ol | 871-91-0 | 99.6% | 126.20 | 100 G
SDP
Catalog No. 5000395949
Encompass_Preferred
7-Octyn-1-ol is a chemical compound that serves as a precursor to 7-Octynoic acid; its oxidation yields 7-Octynoic acid. This compound is also utilized as a click chemistry reagent, characterized by an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing azide groups.
  • Precursor to 7-octynoic acid.
  • Undergoes oxidation to form 7-octynoic acid.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
Catalog No. 50-003-95949 Supplier Medchemexpress LLC Supplier No. HYW008554100G
7-Octyn-1-ol is a chemical compound that serves as a precursor to 7-Octynoic acid; its oxidation yields 7-Octynoic acid. This compound is also utilized as a click chemistry reagent, characterized by an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing azide groups.
  • Precursor to 7-octynoic acid.
  • Undergoes oxidation to form 7-octynoic acid.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.