Learn More
Medchemexpress LLC 7-Octyn-1-ol | 871-91-0 | 99.6% | 126.20 | 25 G
SDP
Catalog No. 5000398805
Encompass_Preferred
7-Octyn-1-ol is a precursor to 7-Octynoic acid, formed through oxidation. It functions as a click chemistry reagent, possessing an Alkyne group that enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Precursor to 7-Octynoic acid
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules
Catalog No. 50-003-98805 Supplier Medchemexpress LLC Supplier No. 25GHYW008554
7-Octyn-1-ol is a precursor to 7-Octynoic acid, formed through oxidation. It functions as a click chemistry reagent, possessing an Alkyne group that enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Precursor to 7-Octynoic acid
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules