Learn More
Medchemexpress LLC 7-Octyn-1-ol | 871-91-0 | 99.6% | 126.20 | 5 G
SDP
Catalog No. 5000422366
Encompass_Preferred
7-Octyn-1-ol is a precursor to 7-Octynoic acid, with its oxidation yielding the acid. This compound functions as a click chemistry reagent, featuring an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules.
  • Precursor to 7-Octynoic acid
  • Click chemistry reagent
  • Contains an alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Liquid appearance
  • Colorless to light yellow
Catalog No. 50-004-22366 Supplier Medchemexpress LLC Supplier No. HYW0085545G
7-Octyn-1-ol is a precursor to 7-Octynoic acid, with its oxidation yielding the acid. This compound functions as a click chemistry reagent, featuring an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules.
  • Precursor to 7-Octynoic acid
  • Click chemistry reagent
  • Contains an alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Liquid appearance
  • Colorless to light yellow