Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Octyn-1-ol | 871-91-0 | 99.6% | 126.20 | 50 G
SDP

Catalog No. 5000423325
Encompass_Preferred

7-Octyn-1-ol is a precursor to 7-Octynoic acid, which is formed through oxidation. It is also a click chemistry reagent containing an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have azide groups.

  • Precursor for 7-octynoic acid
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Purity of 99.60%
  • Molecular weight of 126.20
  • Appears as a colorless to light yellow liquid

Catalog No. 50-004-23325 Supplier Medchemexpress LLC Supplier No. HYW00855450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Octyn-1-ol is a precursor to 7-Octynoic acid, which is formed through oxidation. It is also a click chemistry reagent containing an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have azide groups.

  • Precursor for 7-octynoic acid
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Purity of 99.60%
  • Molecular weight of 126.20
  • Appears as a colorless to light yellow liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.