Learn More
Medchemexpress LLC 7-Octyn-1-ol | 871-91-0 | MFCD01632137 | 99.6% | 126.20 | 10 G
SDP
Catalog No. 5000395805
Encompass_Preferred
7-Octyn-1-ol is a chemical compound with the molecular formula C8H14O. It appears as a colorless to light yellow liquid. This compound serves as a precursor to 7-Octynoic acid, formed via oxidation. It is also utilized as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Precursor to 7-Octynoic acid
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Contains an alkyne group
  • Suitable for use as a click chemistry reagent
Catalog No. 50-003-95805 Supplier Medchemexpress LLC Supplier No. HYW00855410G
7-Octyn-1-ol is a chemical compound with the molecular formula C8H14O. It appears as a colorless to light yellow liquid. This compound serves as a precursor to 7-Octynoic acid, formed via oxidation. It is also utilized as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Precursor to 7-Octynoic acid
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Contains an alkyne group
  • Suitable for use as a click chemistry reagent