Learn More
Medchemexpress LLC 7-Octynoic acid | 10297-09-3 | 98.0% | C8H12O2 | 250 MG
SDP
Catalog No. 5000358684
Encompass_Preferred
7-Octynoic acid is a PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker: one ligand for an E3 ubiquitin ligase and the other for the target protein, leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Click chemistry
  • Alkynes
  • PROTAC synthesis
  • Cancer (disease area)
  • Cancer targeted therapy (disease area)
  • PROTAC
  • PROTAC linkers
Catalog No. 50-003-58684 Supplier Medchemexpress LLC Supplier No. 250MGHY69220
7-Octynoic acid is a PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker: one ligand for an E3 ubiquitin ligase and the other for the target protein, leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Click chemistry
  • Alkynes
  • PROTAC synthesis
  • Cancer (disease area)
  • Cancer targeted therapy (disease area)
  • PROTAC
  • PROTAC linkers